Diphenylcyclopropenone 98%
Glass bottle, 25 g
: 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one
- :886-38-4
- :C₁₅H₁₀O
- :MFCD00001311
- :206.24 g/mol
- :12,03366
- :407 °C (1013 hPa)
- :117…121 °C
- :1,1 g/cm³ (20 °C)
- :Refrigerator
- :212-948-4
- Appearance:Beige to light orange-pink Crystalline powder
- Infrared spectrum:Conforms
- Melting point:117 to 121 °C
- HPLC:≥97.5 %
- Microbial contamination:≤5000 CFU/g (TAMC)
- pH:6.5 to 7.5 (2 % in water)
- Chlorides:≤2 % (as NaCl)
- Total nitrogen:9.3 to 10.7 %
- Amino nitrogen:1.8 to 3.2 %
- Pack type:Glass bottle
- Pk:25 g



