Order Entry
Export
Orders LinkContactUsLinkComponent
Dimetridazole 98%

Dimetridazole 98%

Glass bottle, 100 g
Catalog # ACRO210331000
Irritant
Health
Dimetridazole 98%
Catalog # ACRO210331000
CAS Number:  551-92-8
Dimetridazole 98%
Dimetridazole 98%

Glass bottle, 100 g

CAS Number:  551-92-8
Price
65,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,2-Dimethyl-5-nitroimidazole
  • CAS Number:551-92-8
  • Formula:C₅H₇N₃O₂
  • MDL Number:MFCD00047046
  • Molecular Weight:141.13 g/mol
  • Melting Pt:139…140 °C
  • Storage Temperature:Refrigerator

Documents

Specifications

Specification Test Results

  • Appearance:
    Cream to yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    137 to 142 °C
  • HPLC:
    ≥97.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications