Dimetridazole 98%
Glass bottle, 1 kg
: 1,2-Dimethyl-5-nitroimidazole
- :551-92-8
- :C₅H₇N₃O₂
- :MFCD00047046
- :141.13 g/mol
- :139…140 °C
- :Refrigerator
- Appearance:Cream to yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:137 to 142 °C
- HPLC:≥97.5 %
- Pack type:Glass bottle
- Pk:1 kg



