Order Entry
Export
Orders LinkContactUsLinkComponent
Trypan blue

Trypan blue

Glass bottle, 500 g
Catalog # ACRO189355000
Health
Trypan blue
Catalog # ACRO189355000
CAS Number:  72-57-1
Trypan blue
Trypan blue

Glass bottle, 500 g

CAS Number:  72-57-1
Price
372,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
  • CAS Number:72-57-1
  • Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
  • MDL Number:MFCD00003969
  • Molecular Weight:960.82 g/mol
  • Melting Pt:>280 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-786-7

Documents

Specifications

Specification Test Results

  • Appearance:
    Dark blue to brown-black Powder
  • Infrared spectrum:
    Conforms
  • UV spectrum:
    Complies
  • Dye content:
    ≥60 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications