Sodium peroxide 96%
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: di-Sodium peroxide
- CAS Number:1313-60-6
- Formula:Na₂O₂
- MDL Number:MFCD00003497
- Molecular Weight:77.98 g/mol
- Merck Index:13,08728
- Melting Pt:460 °C (dec.)
- Density:2.8 g/cm³
- Storage Temperature:Ambient
- EINECS:215-209-4
- UN:1504
- ADR:5.1,I
Specifications
Specification Test Results
- Appearance:Yellow Granules (May contain a small quantity of discolored material from the manufacturing process)
- Titration with KMnO4:≥95 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤50 ppm
- Lead (Pb):≤50 ppm
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



