Squalene 99+%
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene
- CAS Number:111-02-4
- Formula:C₃₀H₅₀
- MDL Number:MFCD00008912
- Molecular Weight:410.73 g/mol
- Merck Index:13,08847
- Boiling Pt:285 °C (25 mmHg)
- Melting Pt:–75 °C
- Density:0.855 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:203-826-1
Documents
ACRO20747
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4945 to 1.4965 (20°C, 589 nm)
- Density:(20°C) 0.845 to 0.865 g/ml
- GC:≥99.0 %
- Acid value:≤1 mg KOH/g
- Iodine value:360 to 400 g I2/100 g
- Peroxide value:≤2 meq/kg
- Saponification value:≤1 mg KOH/g
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



