Order Entry
Export
Orders LinkContactUsLinkComponent
cis-4-Hydroxy-D(+)-proline 99%

cis-4-Hydroxy-D(+)-proline 99%

Glass bottle, 10 g
Catalog # ACRO204910100
cis-4-Hydroxy-D(+)-proline 99%
Catalog # ACRO204910100
CAS Number:  2584-71-6
cis-4-Hydroxy-D(+)-proline 99%
cis-4-Hydroxy-D(+)-proline 99%

Glass bottle, 10 g

CAS Number:  2584-71-6
Price
554,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline
  • CAS Number:2584-71-6
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00005252
  • Molecular Weight:131.13 g/mol
  • Melting Pt:243 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:219-963-5

Documents

Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • TLC:
    ≥98.5 %
  • Specific optical rotation:
    +56° to +60° (24°C, 589 nm) (c=2, water)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g

Specifications