Capric acid 99%
Plastic drum, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Decylic acid, Decanoic acid
- CAS Number:334-48-5
- Formula:C₁₀H₂₀O₂
- MDL Number:MFCD00004441
- Molecular Weight:172.27 g/mol
- Merck Index:13,01764
- Boiling Pt:268…270 °C (760 mmHg)
- Melting Pt:30…32 °C
- Density:0.89 g/cm³
- Flash Pt:150 °C
- Storage Temperature:Ambient
- EINECS:206-376-4
Documents
ACRO16727
SDS
Specifications
Specification Test Results
- Appearance:Colorless or white to cream Crystals and/or chunks or fused solid or liquid
- Infrared spectrum:Conforms
- Melting point:29 to 34 °C
- GC:≥98.5 %
Specifications
- Pack type:Plastic drum
- Pk:2,5 kg



