Perylene 99+%
Glass bottle, 5 g
Jump to:
- CAS Number:198-55-0
- Formula:C₂₀H₁₂
- MDL Number:MFCD00004142
- Molecular Weight:252.32 g/mol
- Merck Index:12,07325
- Boiling Pt:443 °C (1013 hPa)
- Melting Pt:277…280 °C
- Density:1,362 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-900-9
Specifications
Specification Test Results
- Appearance:Ochre to brown Powder
- Infrared spectrum:Conforms
- Melting point:277 to 280 °C
- HPLC:≥99.0 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



