Order Entry
Export
Orders LinkContactUsLinkComponent
2-Octanone 99+%

2-Octanone 99+%

Glass bottle, 2,5 L
Catalog # 129420025.
Irritant
Flammable
2-Octanone 99+%
Catalog # 129420025.
CAS Number:  111-13-7
2-Octanone 99+%
2-Octanone 99+%

Glass bottle, 2,5 L

CAS Number:  111-13-7
Price
156,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Octan-2-one
  • CAS Number:111-13-7
  • Formula:C₈H₁₆O
  • MDL Number:MFCD00009540
  • Molecular Weight:128.21 g/mol
  • Merck Index:13,04731
  • Boiling Pt:173 °C (760 mmHg)
  • Melting Pt:–16 °C
  • Density:0.819 g/cm³
  • Flash Pt:55 °C
  • Storage Temperature:Ambient
  • EINECS:203-837-1
  • UN:1224
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4140 to 1.4160 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Clarity of solution:
    Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications