4-(4-Nitrobenzyl)pyridine 98%
Glass bottle, 50 g
Supplier: Thermo Fisher Scientific
4-(4-Nitrobenzyl)pyridine 98%
Catalog # ACRO128550500
Supplier: Thermo Fisher Scientific
CAS Number: 1083-48-3
Jump to:
Product Details & Documents
Synonyms: 4-(p-Nitrobenzyl)Pyridine
- CAS Number:1083-48-3
- Formula:C₁₂H₁₀N₂O₂
- MDL Number:MFCD00006445
- Molecular Weight:214.22 g/mol
- Melting Pt:69…72 °C
- Storage Temperature:Ambient
- EINECS:214-108-2
Documents
ACRO12855
SDS
Specifications
Specification Test Results
- Appearance:Light yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:69 to 72 °C
- Titration with HClO4:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:50 g



