2-Bromopyridine 99%
Glass bottle, 500 mL
- :109-04-6
- :C₅H₄BrN
- :MFCD00006219
- :158 g/mol
- :192…194 °C (761 mmHg)
- :-40,1 °C
- :1.65 g/cm³
- :54 °C
- :Ambient
- :203-641-6
- :2929
- :6.1,II
- Appearance:Clear colorless to yellow to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5700 to 1.5730 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤20 APHA
- Pack type:Glass bottle
- Pk:500 mL



