Order Entry
Export
Orders LinkContactUsLinkComponent
 

Thymol blue, pure indicator

Glass bottle, 5 g
Catalog # ACRO151450050
Thymol blue, pure indicator
Catalog # ACRO151450050
CAS Number:  76-61-9
Thymol blue, pure indicator
Thymol blue, pure indicator

Glass bottle, 5 g

CAS Number:  76-61-9
Price
41,80 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-61-9
  • Formula:C₂₇H₃₀O₅S
  • MDL Number:MFCD00005869
  • Molecular Weight:466.6 g/mol
  • Merck Index:12,09541
  • Melting Pt:221 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-973-3

 

Specifications

Specification Test Results

  • Appearance:
    Dark green to dark brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (0.1 % in ethanol) Clear
  • Specific extinction E (1%/1cm):
    ≥700 (596 nm, in 0.01 N NaOH)
  • Visual transition interval:
    from pH 1.2 (red) to pH 2.8 (yellow)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications