2-Nitrobenzaldehyde 99+%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
2-Nitrobenzaldehyde 99+%
Catalog # ACRO128405000
Supplier: Thermo Fisher Scientific
CAS Number: 552-89-6
Jump to:
Product Details & Documents
- CAS Number:552-89-6
- Formula:C₇H₅NO₃
- MDL Number:MFCD00007132
- Molecular Weight:151.12 g/mol
- Merck Index:12,06684
- Boiling Pt:153 °C (23 mmHg)
- Melting Pt:41…44 °C
- Density:1,35 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:209-025-3
Specifications
Specification Test Results
- Appearance:Yellow Powder or needles and/or chunks
- Infrared spectrum:Conforms
- Melting point:41 to 45 °C
- GC:≥99.0 %
Specifications
- Pack type:Plastic bottle
- Pk:500 g



