Nicotinamide 99%
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
- CAS Number:98-92-0
- Formula:C₆H₆N₂O
- MDL Number:MFCD00006395
- Molecular Weight:122.13 g/mol
- Merck Index:12,06574
- Boiling Pt:150…160 °C (760 mmHg)
- Melting Pt:128…131 °C
- Density:1,4463 g/cm³ (20 °C)
- Flash Pt:>150 °C
- Storage Temperature:Ambient
- EINECS:202-713-4
Documents
ACRO12827
SDS
ACRO128275000_TDS_EN
Product specification
Specifications
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:128 to 131 °C
- Loss on drying:≤0.5 % (4 hrs) (silica gel)
- Sulfated ash:≤0.1 %
- Readily carbonizable substance:Passes test
- pH:6.0 to 7.5 (5 % at 20 °C)
- Titration with HClO4:≥98.5 % (on dried substance)
- Heavy metals (as Pb):≤30 ppm
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



