Order Entry
Export
Orders LinkContactUsLinkComponent
Paracetamol 98%

Paracetamol 98%

Plastic drum, 5 kg
Catalog # ACRO102330051
Irritant
Paracetamol 98%
Catalog # ACRO102330051
CAS Number:  103-90-2
Paracetamol 98%
Paracetamol 98%

Plastic drum, 5 kg

CAS Number:  103-90-2
Price
435,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
  • CAS Number:103-90-2
  • Formula:C₈H₉NO₂
  • MDL Number:MFCD00002328
  • Molecular Weight:151.16 g/mol
  • Merck Index:13,00048
  • Boiling Pt:388 °C (1013 hPa)
  • Melting Pt:168…172 °C
  • Density:1.293 g/cm³
  • Storage Temperature:Ambient
  • EINECS:203-157-5

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    168 to 172 °C
  • HPLC:
    ≥97.5 %
  • Gel strength:
    500 to 1000 g/cm² (1.5 % at 20 °C)
  • Ash:
    ≤6 %
  • Microbial contamination:
    ≤1000 /g (Total plate count)
  • pH:
    5.7 to 7.5 (1.5 % in water)

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 kg

Specifications