Bronopol 98%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropane-1,3-diol
- CAS Number:52-51-7
- Formula:HOCH₂BrC(NO₂)CH₂OH
- MDL Number:MFCD00007390
- Molecular Weight:199.99 g/mol
- Merck Index:13,01432
- Boiling Pt:388 °C (1013 hPa)
- Melting Pt:>120 °C
- Density:1,91 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-143-0
- UN:3241
- ADR:4.1,III
Documents
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:≥120 °C
- Titration Argentometric:≥97.5 % (Ex Br)
- Water:≤0.5 %
- GC:≥97.5 %
- Iron (Fe):≤0.001 %
- Heavy metals (ICP-OES):≤5 ppm
- Sulfur compounds (as SO4):≤0.005 %
- Insoluble matter:≤0.005 %
Specifications
- Pack type:Plastic bottle
- Pk:500 g



