Ethyl 4-hydroxybenzoate 99%
Plastic drum, 5 kg
Supplier: Thermo Fisher Scientific
Ethyl 4-hydroxybenzoate 99%
Catalog # ACRO150250051
Supplier: Thermo Fisher Scientific
CAS Number: 120-47-8
Jump to:
Product Details & Documents
Synonyms: Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
- CAS Number:120-47-8
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00002353
- Molecular Weight:166.18 g/mol
- Merck Index:13,03869
- Boiling Pt:297…298 °C (760 mmHg)
- Melting Pt:115…118 °C
- Density:1,078 g/cm³ (131 °C)
- Storage Temperature:Ambient
- EINECS:204-399-4
Documents
Specifications
Specification Test Results
- Appearance:White to light cream Crystalline powder
- Infrared spectrum:Conforms
- Melting point:115 to 118 °C
- GC:≥98.5 %
Specifications
- Pack type:Plastic drum
- Pk:5 kg



