Ethyl propiolate 99%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Ethyl propynoate, 2-Propynoic acid ethyl ester, Ethyl acetylenecarboxylate, Propiolic acid ethyl ester
- CAS Number:623-47-2
- Formula:C₅H₆O₂
- MDL Number:MFCD00009184
- Molecular Weight:98.1 g/mol
- Boiling Pt:120 °C (760 mmHg)
- Melting Pt:9 °C
- Density:0.968 g/cm³
- Flash Pt:23 °C
- Storage Temperature:Refrigerator
- EINECS:210-795-8
- UN:1993
- ADR:3,III
Documents
ACRO11890
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4110 to 1.4130 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:100 g



