Acetaldehyde 99.5%, extra pure
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Acetaldehyde 99.5%, extra pure
Catalog # 149510025.
Supplier: Thermo Fisher Scientific
CAS Number: 75-07-0
Jump to:
Product Details & Documents
Synonyms: Ethanal
- CAS Number:75-07-0
- Formula:CH₃CHO
- MDL Number:MFCD00006991
- Molecular Weight:44.05 g/mol
- Merck Index:12,00037
- Boiling Pt:21 °C (760 mmHg)
- Melting Pt:–123 °C
- Density:0.785 g/cm³
- Flash Pt:–27 °C
- Storage Temperature:Refrigerator
- EINECS:200-836-8
- UN:1089
- ADR:3,I
Documents
Certificates
Search by Lot NumberACRO14951
SDS
ACRO149510025_Specification sheet.pdf
Product specification
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3310 to 1.3330 (20°C, 589 nm)
- Water:≤0.02 %
- GC:≥99.4 %
- Chlorine (Cl):≤0.003 % (total)
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



