Order Entry
Export
Orders LinkContactUsLinkComponent
Phenol 99%, extra pure

Phenol 99%, extra pure

Plastic drum, 5 kg
Catalog # 149340051.
Corrosive
Toxic
Health
Environment
Phenol 99%, extra pure
Catalog # 149340051.
CAS Number:  108-95-2
Phenol 99%, extra pure
Phenol 99%, extra pure

Plastic drum, 5 kg

CAS Number:  108-95-2
Price
200,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Merck Index:12,07390
  • Boiling Pt:182 °C (760 mmHg)
  • Melting Pt:39…42 °C
  • Density:1.07 g/cm³
  • Flash Pt:79 °C
  • Storage Temperature:Ambient
  • EINECS:203-632-7
  • UN:1671
  • ADR:6.1,II
  • REACH:01-2119471329-32

Documents

Specifications

Specification Test Results

  • Appearance:
    Colorless to white Crystals or fused mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    39 to 42 °C
  • Water:
    ≤0.2 %
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    5 kg

Specifications