1,2-Ethanedithiol 95%
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
1,2-Ethanedithiol 95%
Catalog # ACRO117850010
Supplier: Thermo Fisher Scientific
CAS Number: 540-63-6
Jump to:
Product Details & Documents
Synonyms: 1.2-Ethanedithiol, 1,2-Dimercaptoethane
- CAS Number:540-63-6
- Formula:C₂H₆S₂
- MDL Number:MFCD00004892
- Molecular Weight:94.2 g/mol
- Merck Index:13,03760
- Boiling Pt:144…146 °C (760 mmHg)
- Melting Pt:–41 °C
- Density:1.123 g/cm³
- Flash Pt:50 °C
- Storage Temperature:Ambient
- EINECS:208-752-3
- UN:3071
- ADR:6.1,II
Documents
ACRO11785
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow to slightly colored Liquid
- Infrared spectrum:Conforms
- GC:≥94 %
- Bromide (Br):≤0.01 %
- Iodide (I):≤20 ppm
- Chlorate and Nitrate (as NO3):≤0.003 %
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤10 ppm
- Barium (Ba):Passes test (Limit about 0.001%)
- Calcium (Ca):≤20 ppm
- Iron (Fe):≤3 ppm
- Magnesium (Mg):≤10 ppm
- Sodium (Na):≤50 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 %
Specifications
- Pack type:Glass bottle
- Pk:1 L



