n-Dodecane 99%, pure
Metal drum, 10 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Dodecane
- CAS Number:112-40-3
- Formula:H₃C(CH₂)₁₀CH₃
- MDL Number:MFCD00008969
- Molecular Weight:170.34 g/mol
- Boiling Pt:215…217 °C (760 mmHg)
- Melting Pt:–12 °C
- Density:0.753 g/cm³
- Flash Pt:71 °C
- Storage Temperature:Ambient
- EINECS:203-967-9
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4210 to 1.4230 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤10 APHA
- Heavy metals (ICP-OES):≤5 ppm (as Pb)
- Titrable acid:≤0.0008 meq/g
- Insoluble matter:≤0.01 %
- Reducing sugars:Passes test
Specifications
- Pack type:Metal drum
- Pk:10 L



