(±)-Mandelic acid 99+%
Plastic drum, 5 kg
: DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid
- :90-64-2
- :C₈H₈O₃
- :MFCD00064250
- :152.15 g/mol
- :322 °C (1013 hPa)
- :118…122 °C
- :1,361 g/cm³ (20 °C)
- :Ambient
- :210-277-1
Documents
ACRO12531
SDS
ACRO125310010_TDS_EN
Product specification
- Appearance:White to light yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:117 to 121 °C
- Titration with NaOH:≥99.0 %
- Nitrate (NO3):≤0.001 %
- Phosphate (PO4):≤0.0003 %
- Aluminium (Al):≤0.001 %
- Arsenic (As):≤0.00002 %
- Cadmium (Cd):≤0.0005 %
- Calcium (Ca):≤0.001 %
- Chromium (Cr):≤0.0005 %
- Cobalt (Co):≤0.0005 %
- Copper (Cu):≤0.0005 %
- Iron (Fe):≤0.002 %
- Lead (Pb):≤0.0005 %
- Magnesium (Mg):≤0.0005 %
- Manganese (Mn):≤0.0005 %
- Nickel (Ni):≤0.0005 %
- Potassium (K):≤0.01 %
- Sodium (Na):≤0.005 %
- Zinc (Zn):≤0.0005 %
- Heavy metals (as Pb):≤20 ppm
- Insoluble matter:≤0.005 % (in water)
- Pack type:Plastic drum
- Pk:5 kg



