2,6-Lutidine 99%
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 2,6-Dimethylpyridine
- CAS Number:108-48-5
- Formula:C₇H₉N
- MDL Number:MFCD00006345
- Molecular Weight:107.16 g/mol
- Merck Index:13,05636
- Boiling Pt:143…145 °C (760 mmHg)
- Melting Pt:–6 °C
- Density:0.92 g/cm³
- Flash Pt:33 °C
- Storage Temperature:Ambient
- EINECS:203-587-3
- UN:1993
- ADR:3,III
Documents
ACRO12519
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4966 to 1.4986 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



