Bathophenanthroline 99%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Bathophenanthroline 99%
Catalog # ACRO158320250
Supplier: Thermo Fisher Scientific
CAS Number: 1662-01-7
Jump to:
Product Details & Documents
Synonyms: 4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)
- CAS Number:1662-01-7
- Formula:C₂₄H₁₆N₂
- MDL Number:MFCD00004976
- Molecular Weight:332.4 g/mol
- Boiling Pt:508 °C (1013 hPa)
- Melting Pt:218…221 °C
- Storage Temperature:Ambient
- EINECS:216-767-1
Specifications
Specification Test Results
- Appearance:White to cream to light yellow Powder
- Infrared spectrum:Conforms
- Melting point:218 to 222 °C
- Titration with HClO4:≥98.5 %
- Suitability:(for Fe determination) Passes test
- UV:at 280 nm A: ≤0.01
- Color scale:≤15 APHA (0.1 M in water)
- pH:5.0 to 9.0
- Heavy metals (as Pb):≤0.001 %
Specifications
- Pack type:Glass bottle
- Pk:25 g



