3,5-Dimethoxyphenol 97%
Glass bottle, 50 g
Supplier: Thermo Fisher Scientific
3,5-Dimethoxyphenol 97%
Catalog # ACRO158120500
Supplier: Thermo Fisher Scientific
CAS Number: 500-99-2
Jump to:
Product Details & Documents
- CAS Number:500-99-2
- Formula:C₈H₁₀O₃
- MDL Number:MFCD00008388
- Molecular Weight:154.17 g/mol
- Boiling Pt:172…175 °C (17 mmHg)
- Melting Pt:40…45 °C
- Flash Pt:78 °C
- Storage Temperature:Ambient
- EINECS:207-917-7
Documents
ACRO15812
SDS
Specifications
Specification Test Results
- Appearance:White to yellow to pink Crystalline powder or crystals or crystalline mass
- Infrared spectrum:Conforms
- Melting point:40 to 46 °C
- GC:≥96.0 %
- UV:at 280 nm A: ≤0.01
- pH:6.5 to 7.3 (5 % in water)
- Titration with HClO4:≥97.5 %
- Heavy metals (as Pb):≤2 ppm
Specifications
- Pack type:Glass bottle
- Pk:50 g



