2,5-Dimethoxybenzaldehyde 97%
Glass bottle, 500 g
Supplier: Thermo Fisher Scientific
2,5-Dimethoxybenzaldehyde 97%
Catalog # ACRO115395000
Supplier: Thermo Fisher Scientific
CAS Number: 93-02-7
Jump to:
Product Details & Documents
- CAS Number:93-02-7
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00003314
- Molecular Weight:166.18 g/mol
- Boiling Pt:146 °C (10 mmHg)
- Melting Pt:45…50 °C
- Flash Pt:113 °C
- Storage Temperature:Ambient
- EINECS:202-211-5
Documents
ACRO11539
SDS
Specifications
Specification Test Results
- Appearance:Yellow to beige Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:46 to 51 °C
- Water:≤0.5 %
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:500 g



