2,7-Dihydroxynaphthalene 97%
Glass bottle, 500 g
Supplier: Thermo Fisher Scientific
2,7-Dihydroxynaphthalene 97%
Catalog # ACRO115045000
Supplier: Thermo Fisher Scientific
CAS Number: 582-17-2
Jump to:
Product Details & Documents
Synonyms: 2,7-Naphthalenediol, Naphthalene-2,7-diol
- CAS Number:582-17-2
- Formula:C₁₀H₈O₂
- MDL Number:MFCD00004085
- Molecular Weight:160.17 g/mol
- Boiling Pt:375 °C (1013 hPa)
- Melting Pt:184…189 °C
- Storage Temperature:Ambient
- EINECS:209-478-7
Documents
Specifications
Specification Test Results
- Appearance:Grey Powder
- Infrared spectrum:Conforms
- Melting point:184 to 189 °C
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:500 g



