Order Entry
Export
Orders LinkContactUsLinkComponent
2,5-Dihydroxybenzaldehyde 99%

2,5-Dihydroxybenzaldehyde 99%

Glass bottle, 25 g
Catalog # ACRO114840250
Irritant
2,5-Dihydroxybenzaldehyde 99%
Catalog # ACRO114840250
CAS Number:  1194-98-5
2,5-Dihydroxybenzaldehyde 99%
2,5-Dihydroxybenzaldehyde 99%

Glass bottle, 25 g

CAS Number:  1194-98-5
Price
255,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:1194-98-5
  • Formula:C₇H₆O₃
  • MDL Number:MFCD00003333
  • Molecular Weight:138.12 g/mol
  • Boiling Pt:60 - 65 °C (12 -rr)
  • Melting Pt:98…102 °C
  • Density:0,859 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:214-789-6

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow to khaki Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    98 to 102 °C
  • GC:
    ≥98.5 %
  • Copper (Cu):
    ≤50 ppm
  • Iron (Fe):
    ≤100 ppm
  • Lead (Pb):
    ≤50 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications