Order Entry
Export
Orders LinkContactUsLinkComponent
 

D-(+)-Xylose 99+%

Plastic bottle, 2,5 kg
Catalog # ACRO141000025
D-(+)-Xylose 99+%
Catalog # ACRO141000025
CAS Number:  58-86-6
D-(+)-Xylose 99+%
D-(+)-Xylose 99+%

Plastic bottle, 2,5 kg

CAS Number:  58-86-6
Price
801,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
  • CAS Number:58-86-6
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00151475
  • Molecular Weight:150.13 g/mol
  • Merck Index:13,10142
  • Boiling Pt:331 °C (1013 hPa)
  • Melting Pt:148…153 °C
  • Density:1,525 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-400-7

 

Specifications

Specification Test Results

  • Appearance:
    White to almost white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.2 % (K.F.)
  • Sulfated ash:
    ≤0.2 %
  • HPLC:
    ≥99.0 % (ELSD)
  • Specific optical rotation:
    +18.0° to +21.0° (20°C, 589 nm) (c=10, H2O/NH3)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg

Specifications