Diethyl phthalate 99%
Plastic drum, 10 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Phthalic acid diethyl ester
- CAS Number:84-66-2
- Formula:C₁₂H₁₄O₄
- MDL Number:MFCD00009111
- Molecular Weight:222.24 g/mol
- Merck Index:12,07527
- Boiling Pt:298…299 °C (760 mmHg)
- Melting Pt:-3 °C
- Density:1.118 g/cm³
- Flash Pt:156 °C
- Storage Temperature:Ambient
- EINECS:201-550-6
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5010 to 1.5030 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥98.5 %
- Color scale:≤25 APHA
Specifications
- Pack type:Plastic drum
- Pk:10 L



