4-Nitrophenol 99%
Plastic drum, 5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1-Hydroxy-4-nitrobenzene, p-Nitrophenol
- CAS Number:100-02-7
- Formula:C₆H₅NO₃
- MDL Number:MFCD00007331
- Molecular Weight:139.11 g/mol
- Merck Index:13,06654
- Boiling Pt:279 °C (760 mmHg)
- Melting Pt:>112 °C
- Density:1,48 g/cm³ (20 °C)
- Flash Pt:169 °C
- Storage Temperature:Ambient
- EINECS:202-811-7
- UN:1663
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:Yellow to brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:112 to 115 °C
- GC:≥98.5 %
- Titration with NaOH:≥96.0 to ≤104.0 %
Specifications
- Pack type:Plastic drum
- Pk:5 kg



