n-Undecane 99%
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Undecane
- CAS Number:1120-21-4
- Formula:H₃C(CH₂)₉CH₃
- MDL Number:MFCD00008959
- Molecular Weight:156.31 g/mol
- Boiling Pt:196 °C (760 mmHg)
- Melting Pt:–26 °C
- Density:0.74 g/cm³
- Flash Pt:65 °C
- Storage Temperature:Ambient
- EINECS:214-300-6
- UN:2330
- ADR:3,III
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4160 to 1.4180 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



