Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,2-Dibromoethane 99%

Plastic bottle, 2,5 L
Catalog # ACRO112790025
Toxic
Health
Environment
1,2-Dibromoethane 99%
Catalog # ACRO112790025
CAS Number:  106-93-4
1,2-Dibromoethane 99%
1,2-Dibromoethane 99%

Plastic bottle, 2,5 L

CAS Number:  106-93-4
Price
264,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
  • CAS Number:106-93-4
  • Formula:BrH₂CCH₂Br
  • MDL Number:MFCD00000233
  • Molecular Weight:187.86 g/mol
  • Merck Index:13,03830
  • Boiling Pt:131…132°C (760mmHg)
  • Boiling Pt:131…132 °C (760 mmHg)
  • Melting Pt:9…10°C
  • Melting Pt:9…10 °C
  • Density:2.173
  • Flash Pt:>104°C
  • Flash Pt:>104 °C
  • Storage Temperature:Ambient
  • EINECS:203-444-5
  • UN:1605
  • ADR:6.1,I
  • ADR:6.1,

 

Specifications

Specification Test Results

  • Appearance:
    clear colorless to light yellow liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥98.5%
  • Refractive index:
    1.5375 to 1.5395 (20°C, 589nm)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L

Specifications