1,3-Dibromopropane 98% stabilised
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
1,3-Dibromopropane 98% stabilised
Catalog # ACRO155040025
Supplier: Thermo Fisher Scientific
CAS Number: 109-64-8
Jump to:
Product Details & Documents
- CAS Number:109-64-8
- Formula:BrH₂CCH₂CH₂Br
- MDL Number:MFCD00000255
- Molecular Weight:201.89 g/mol
- Merck Index:12,09844
- Boiling Pt:167 °C (760 mmHg)
- Melting Pt:–34 °C
- Density:1.989 g/cm³
- Flash Pt:54 °C
- Storage Temperature:Ambient
- EINECS:203-690-3
- UN:1993
- ADR:3,III
Documents
ACRO15504
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5220 to 1.5240 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:400 ppm butylene oxide
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



