2,3-Diaminopyridine 98%
Glass bottle, 100 g
: Pyridine-2,3-diyldiamine
- :452-58-4
- :C₅H₇N₃
- :MFCD00006319
- :109.13 g/mol
- :148…150 °C (5 mmHg)
- :114…116 °C
- :205 °C
- :Refrigerator
- :207-200-9
- :2811
- :6.1,II
- Appearance:Grey to dark brown Crystalline powder or needles (May darken on storage)
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Titration with HClO4:97.5 to 102.5 %
- NMR (1H-NMR):Consistent with structure
- Pack type:Glass bottle
- Pk:100 g



