Cyclohexene 99% stabilised, pure
Plastic drum, 10 L
Supplier: Thermo Fisher Scientific
Cyclohexene 99% stabilised, pure
Catalog # ACRO154840100
Supplier: Thermo Fisher Scientific
CAS Number: 110-83-8
Jump to:
Product Details & Documents
- CAS Number:110-83-8
- Formula:C₆H₁₀
- MDL Number:MFCD00001539
- Molecular Weight:82.15 g/mol
- Merck Index:13,02756
- Boiling Pt:83 °C (760 mmHg)
- Melting Pt:–104 °C
- Density:0.81 g/cm³
- Flash Pt:–20 °C
- Storage Temperature:Ambient
- EINECS:203-807-8
- UN:2256
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4450 to 1.4480 (20°C, 589 nm)
- GC:≥98.5 %
- Stabilizer:Approx. 100 ppm 2-tert-Butyl-4-methylphenol
Specifications
- Pack type:Plastic drum
- Pk:10 L



