Order Entry
Export
Orders LinkContactUsLinkComponent
2,4,6-Trichlorophenol 98%

2,4,6-Trichlorophenol 98%

Glass bottle, 500 g
Catalog # ACRO139485000
Irritant
Health
Environment
2,4,6-Trichlorophenol 98%
Catalog # ACRO139485000
CAS Number:  88-06-2
2,4,6-Trichlorophenol 98%
2,4,6-Trichlorophenol 98%

Glass bottle, 500 g

CAS Number:  88-06-2
Price
97,30 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:88-06-2
  • Formula:C₆H₃Cl₃O
  • MDL Number:MFCD00002172
  • Molecular Weight:197.45 g/mol
  • Merck Index:12,09773
  • Boiling Pt:246 °C (760 mmHg)
  • Melting Pt:64…68 °C
  • Density:1,675 g/cm³ (25 °C)
  • Flash Pt:99 °C
  • Storage Temperature:Ambient
  • EINECS:201-795-9
  • UN:2020
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Light beige to light brown Crystalline powder or mass and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    64 to 68 °C
  • GC:
    ≥97.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications