2'-Hydroxy-4'-methoxyacetophenone 99%
Glass bottle, 50 g
: 1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
- :552-41-0
- :C₉H₁₀O₃
- :MFCD00008730
- :166.18 g/mol
- :154 °C (20 -rr)
- :47…50 °C
- :1,1604 g/cm³ (40 °C)
- :>110 °C
- :Ambient
- :209-012-2
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:47 to 50 °C
- GC:≥98.5 %
- Stabilizer:Approx. 100 ppm Amylene
- Residue after evaporation:≤0.002 %
- Pack type:Glass bottle
- Pk:50 g



