1,3-Diaminopropane 99%
Glass bottle, 50 mL
Supplier: Thermo Fisher Scientific
1,3-Diaminopropane 99%
Catalog # ACRO112350500
Supplier: Thermo Fisher Scientific
CAS Number: 109-76-2
Jump to:
Product Details & Documents
Synonyms: 1,3-Propanediamine, Trimethylenediamine
- CAS Number:109-76-2
- Formula:C₃H₁₀N₂
- MDL Number:MFCD00008228
- Molecular Weight:74.13 g/mol
- Boiling Pt:140 °C (760 mmHg)
- Melting Pt:–12 °C
- Density:0.88 g/cm³
- Flash Pt:48 °C
- Storage Temperature:Ambient
- EINECS:203-702-7
- UN:2734
- ADR:8,I
Documents
ACRO11235
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4565 to 1.4585 (20°C, 589 nm)
- Water:≤0.5 %
- GC:≥98.5 %
- Color scale:≤20 APHA
- Titration with HClO4:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:50 mL



