Order Entry
Export
Orders LinkContactUsLinkComponent
L-Threonine 98%

L-Threonine 98%

Glass bottle, 5 g
Catalog # ACRO138930050
L-Threonine 98%
Catalog # ACRO138930050
CAS Number:  72-19-5
L-Threonine 98%
L-Threonine 98%

Glass bottle, 5 g

CAS Number:  72-19-5
Price
22,80 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
  • CAS Number:72-19-5
  • Formula:C₄H₉NO₃
  • MDL Number:MFCD00064270
  • Molecular Weight:119.12 g/mol
  • Melting Pt:255 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-774-1

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or glistening flakes
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 % (1 g, 105°C, 3 hrs)
  • Sulfated ash:
    ≤0.1 %
  • Solubility:
    (2.5 % in water) Clear colorless
  • Specific optical rotation:
    -27.6° to -29.0° (20°C, 589 nm) (c=6, H2O)
  • Titration with HClO4:
    97.5 to 102.5 % (On dried substance)
  • Chloride (Cl):
    ≤200 ppm
  • Sulfate (SO4):
    ≤300 ppm
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications