3,3'-Thiodipropionic acid 99%
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
3,3'-Thiodipropionic acid 99%
Catalog # ACRO138750025
Supplier: Thermo Fisher Scientific
CAS Number: 111-17-1
Jump to:
Product Details & Documents
Synonyms: 3,3-Thiodipropionic acid
- CAS Number:111-17-1
- Formula:C₆H₁₀O₄S
- MDL Number:MFCD00002781
- Molecular Weight:178.21 g/mol
- Merck Index:12,09468
- Boiling Pt:347 °C (1013 hPa)
- Melting Pt:130…133 °C
- Density:1,496 g/cm³ (20 °C)
- Flash Pt:128 °C
- Storage Temperature:Ambient
- EINECS:203-841-3
Documents
ACRO13875
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:130 to 133 °C
- Titration with NaOH:98.5 to 101.5 %
- NMR (1H-NMR):Consistent with structure
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



