3,3-Dimethylpentane 99%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
3,3-Dimethylpentane 99%
Catalog # ACRO152660250
Supplier: Thermo Fisher Scientific
CAS Number: 562-49-2
Jump to:
Product Details & Documents
- CAS Number:562-49-2
- Formula:C₇H₁₆
- MDL Number:MFCD00009322
- Molecular Weight:100.2 g/mol
- Boiling Pt:86 °C (760 mmHg)
- Melting Pt:–135 °C
- Density:0.69 g/cm³
- Flash Pt:–6 °C
- Storage Temperature:Ambient
- EINECS:209-230-8
- ADR:3,II
- :Density: 0.6936 g/cm³ (20 °C)
- UN:1206
Documents
ACRO15266
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3900 to 1.3920 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:25 g



