Order Entry
Export
Orders LinkContactUsLinkComponent
m-Cresol 99%

m-Cresol 99%

Glass bottle, 100 g
Catalog # ACRO110581000
Toxic
Corrosive
m-Cresol 99%
Catalog # ACRO110581000
CAS Number:  108-39-4
m-Cresol 99%
m-Cresol 99%

Glass bottle, 100 g

CAS Number:  108-39-4
Price
38,10 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-Hydroxy-3-methylbenzene, 3-Methylphenol, 3-Hydroxytoluol, 3-Hydroxytoluene
  • CAS Number:108-39-4
  • Formula:CH₃C₆H₄OH
  • MDL Number:MFCD00002302
  • Molecular Weight:108.14 g/mol
  • Merck Index:13,02604
  • Boiling Pt:203 °C (760 mmHg)
  • Melting Pt:8…10 °C
  • Density:1.03 g/cm³
  • Flash Pt:86 °C
  • Storage Temperature:Ambient
  • EINECS:203-577-9
  • UN:2076
  • ADR:6.1,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light red Viscous liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5390 to 1.5420 (20°C, 589 nm)
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g

Specifications