Potassium hydroxide ∼ 85%, pellets for analysis
Fibre drum, 25 kg
Supplier: Thermo Fisher Scientific
Potassium hydroxide ∼ 85%, pellets for analysis
Catalog # ACRO134060250
Supplier: Thermo Fisher Scientific
CAS Number: 1310-58-3
Jump to:
Product Details & Documents
Synonyms: Potash caustic, Caustic potash
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Merck Index:12,07806
- Boiling Pt:1320 °C (760 mmHg)
- Melting Pt:360 °C
- Density:2,04 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-181-3
- UN:1813
- ADR:8,II
- REACH:01-2119487136-33
Documents
Specifications
Specification Test Results
- Appearance:White Pellets
- Titration with HCl:≥85 %
- Chloride (Cl):≤70 ppm
- Phosphate (PO4):≤4 ppm
- Sulfate (SO4):≤10 ppm
- Potassium carbonate:≤1 % (K2CO3)
- Aluminium (Al):≤10 ppm
- Arsenic (As):≤0.5 ppm
- Calcium (Ca):≤10 ppm
- Iron (Fe):≤5 ppm
- Nickel (Ni):≤3 ppm
- Silver (Ag):≤0.5 ppm
- Sodium (Na):≤0.4 %
- Heavy metals (as Pb):≤10 ppm
- Silicon dioxide (SiO2):≤5 ppm
- Nitrogen compounds (as N):≤5 ppm
Specifications
- Pack type:Fibre drum
- Pk:25 kg



