Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Propanedithiol 98%

1,3-Propanedithiol 98%

Glass bottle, 250 mL
Catalog # ACRO131412500
Flammable
Toxic
1,3-Propanedithiol 98%
Catalog # ACRO131412500
CAS Number:  109-80-8
1,3-Propanedithiol 98%
1,3-Propanedithiol 98%

Glass bottle, 250 mL

CAS Number:  109-80-8
Price
292,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  1,3-Dimercaptopropane
  • CAS Number:109-80-8
  • Formula:C₃H₈S₂
  • MDL Number:MFCD00004904
  • Molecular Weight:108.23 g/mol
  • Merck Index:12,07984
  • Boiling Pt:169 °C (760 mmHg)
  • Melting Pt:–79 °C
  • Density:1.07 g/cm³
  • Flash Pt:58 °C
  • Storage Temperature:Ambient
  • EINECS:203-706-9
  • UN:1228
  • ADR:3,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5380 to 1.5420 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Density:
    (20°C) 1.038 to 1.048 g/ml

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications