1,3-Propanediol 98%
Metal drum, 10 L
1,3-Propanediol 98%
ACRO131400100
: 504-63-2
: 1,3-Propylene glycol, 1,3-Dihydroxy propane
- :504-63-2
- :HO(CH₂)₃OH
- :MFCD00002949
- :76.1 g/mol
- :12,09845
- :214 °C (760 mmHg)
- :–32 °C
- :1.052 g/cm³
- :140 °C
- :Ambient
- :207-997-3
- Appearance:Clear viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4380 to 1.4400 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥97.5 %
- Color scale:≤20 APHA
- UV:at 280 nm A: ≤0.301
- Residue after evaporation:≤0.001 %
- Pack type:Metal drum
- Pk:10 L
- Cat. No.:ACRO131400100
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating)
- Environmentally Preferable:Safer Human and Environmental Health



