Order Entry
Export
Orders LinkContactUsLinkComponent
Pepsin

Pepsin

Glass bottle, 20 g
Catalog # ACRO417070200
Pepsin
Catalog # ACRO417070200
CAS Number:  
Pepsin

Glass bottle, 20 g

CAS Number:  
Price
41,70 €
Log in for availability
1 * 20 g
Product Details & Documents

Contains either Lactose or Maltodextrin excipient

  • Assay: >=0.5 Ph. Eur. U/mg (Activity)
  • Assay: Hemoglobin, pH 1.6, 25°C
  • Infrared spectrum: Conforms
  • Loss on drying: =<5.0 % (60°C, 4 h, 670 Pa) (P2O5)
  • Microbial contamination: Passes test
  • Sulfated ash: =<5.0 % (800°C)

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    20 g
  • Cat. No.:
    ACRO417070200
  • Form:
    White to light yellow powder
  • Storage conditions:
    Refrigerator
  • pH:
    3.0 to 5.0 (2 % in water)
  • MDL No.:
    MFCD00081840
  • Enzyme name:
    Pepsin
  • CAS No.:
    9001-75-6

Specifications