Order Entry
Export
Orders LinkContactUsLinkComponent
Pepsin

Pepsin

Plastic bottle, 2,5 kg
  ACRO417070025
Pepsin
  ACRO417070025
 :  
Pepsin

Plastic bottle, 2,5 kg

 :  
 
1 550,00 €
   
1 * 2,5 kg
 

Contains either Lactose or Maltodextrin excipient

  • Assay: >=0.5 Ph. Eur. U/mg (Activity)
  • Assay: Hemoglobin, pH 1.6, 25°C
  • Infrared spectrum: Conforms
  • Loss on drying: =<5.0 % (60°C, 4 h, 670 Pa) (P2O5)
  • Microbial contamination: Passes test
  • Sulfated ash: =<5.0 % (800°C)

Documents

 

 

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg
  • Cat. No.:
    ACRO417070025
  • Form:
    White to light yellow powder
  • Storage conditions:
    Refrigerator
  • pH:
    3.0 to 5.0 (2 % in water)
  • MDL No.:
    MFCD00081840
  • Enzyme name:
    Pepsin
  • CAS No.:
    9001-75-6